ADV465748325 AChemBlock

(3R,4S)-4-Amino-tetrahydropyran-3-ol hydrochloride

  • MDL number MFCD23159220
  • Cas Number : 1096594-11-4
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3r-4s-4-amino-tetrahydropyran-3-ol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9259
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart