ADV465748326 AChemBlock

cis-3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]cyclobutanemethanol

  • MDL number MFCD23106335
  • Cas Number : 959749-93-0
  • Molecular Weight : 216.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/cis-3-1-1-dimethylethyl-dimethylsilyl-oxy-cyclobutanemethanol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9260
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart