ADV465748332 AChemBlock

trans-3-N-Boc-3-amino-4-hydroxy-tetrahydrofuran

  • MDL number MFCD11617817
  • Cas Number : 1430230-65-1
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/trans-3-n-boc-3-amino-4-hydroxy-tetrahydrofuran.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9266
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart