ADV465748334 AChemBlock

3-({[(tert-Butoxy)carbonyl]amino}methyl)cyclobutane-1-carboxylic acid

  • MDL number MFCD27987241
  • Cas Number : 1630907-04-8
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-tert-butoxy-carbonyl-amino-methyl-cyclobutane-1-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9268
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart