ADV465748340 AChemBlock

2-Methyl-5-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,4-oxadiazole

  • MDL number MFCD18384931
  • Cas Number : 1056456-24-6
  • Molecular Weight : 286.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-5-4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-phenyl-1-3-4-oxadiazole.html

Safety Information

signalword Warning
pcodes P264;P270;P301+312+330;P502
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9274
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart