ADV465748342 AChemBlock

1H-Imidazole-2-carbonitrile hydrochloride

  • MDL number MFCD22573391
  • Cas Number : 1392803-62-1
  • Molecular Weight : 129.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1h-imidazole-2-carbonitrile-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2923-01
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart