ADV465748347 AChemBlock

cis-3-Aminocyclohexane-1-carboxylic acid hydrochloride

  • MDL number MFCD00787794
  • Cas Number : 118785-96-9
  • Molecular Weight : 179.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/cis-3-aminocyclohexane-1-carboxylic-acid-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9281
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart