ADV465748352 AChemBlock

(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-hydroxymethylpyrrolidine

  • MDL number MFCD22573266
  • Cas Number : 1138324-48-7
  • Molecular Weight : 219.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2s-4r-1-tert-butoxycarbonyl-4-fluoro-2-hydroxymethylpyrrolidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9286
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart