ADV465748354 AChemBlock

2,2,2-Trifluoro-1-(pyridin-2-yl)ethanamine hydrochloride

  • MDL number MFCD11506260
  • Cas Number : 1187929-38-9
  • Molecular Weight : 212.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-2-2-trifluoro-1-pyridin-2-yl-ethanamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9288
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart