ADV465748370 AChemBlock

3-(Iodomethyl)tetrahydrofuran

  • MDL number MFCD06410690
  • Cas Number : 475060-43-6
  • Molecular Weight : 212.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Light yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-iodomethyl-tetrahydrofuran.html
Availability details for
Quantity
  • Availability
  • Add to Cart