ADV465748374 AChemBlock

tert-Butyl N-(trans-3-aminocyclobutyl)-N-methylcarbamate

  • MDL number MFCD23106178
  • Cas Number : 1392803-14-3
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-trans-3-aminocyclobutyl-n-methylcarbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9308
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart