ADV465748375 AChemBlock

Methyl 2-(benzyloxycarbonylamino)-2-(oxetan-3-ylidene)acetate

  • MDL number MFCD14586429
  • Cas Number : 394653-39-5
  • Molecular Weight : 277.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-benzyloxycarbonylamino-2-oxetan-3-ylidene-acetate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9309
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart