ADV465748376 AChemBlock

3,3-Dimethylpyrrolidine hydrochloride

  • MDL number MFCD11849606
  • Cas Number : 792915-20-9
  • Molecular Weight : 135.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-3-dimethylpyrrolidine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9310
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart