ADV465748388 AChemBlock

tert-Butyl (2S,4R)-2-(aminomethyl)-4-fluoro-1-pyrrolidinecarboxylate

  • MDL number MFCD23106393
  • Cas Number : 1138324-46-5
  • Molecular Weight : 218.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2s-4r-2-aminomethyl-4-fluoro-1-pyrrolidinecarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9323
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart