ADV465748398 AChemBlock

4-Chloro-3-iodo-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD17016016
  • Cas Number : 949558-30-9
  • Molecular Weight : 279.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-3-iodo-1h-pyrazolo-3-4-b-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9333
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart