ADV465748400 AChemBlock

6-tert-Butyl-4-chlorothieno[3,2-d]pyrimidine

  • MDL number MFCD03407483
  • Cas Number : 439693-52-4
  • Molecular Weight : 226.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/6-tert-butyl-4-chlorothieno-3-2-d-pyrimidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9335
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart