ADV465748403 AChemBlock

ethyl 5-chloropyrimidine-2-carboxylate

  • MDL number MFCD16610953
  • Cas Number : 1227571-82-5
  • Molecular Weight : 186.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-pyrimidinecarboxylic-acid-5-chloro-ethyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9339
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart