ADV465748407 AChemBlock

8-Bromo-4-chloro-7-methoxy-1,6-naphthyridine

  • MDL number MFCD27956900
  • Cas Number : 1434142-06-9
  • Molecular Weight : 273.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/8-bromo-4-chloro-7-methoxy-1-6-naphthyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9343
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart