ADV465748414 AChemBlock

2-Boc-7-formyl-1,2,3,4-tetrahydropyrrolo[3,2,1-jk][1,4]benzodiazepine

  • MDL number MFCD21642014
  • Cas Number : 1122597-86-7
  • Molecular Weight : 300.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-boc-7-formyl-1-2-3-4-tetrahydropyrrolo-3-2-1-jk-1-4-benzodiazepine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9350
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart