ADV465748420 AChemBlock

tert-Butyl ([4-(hydroxymethyl)-4-piperidinyl]methyl)carbamate

  • MDL number MFCD22397604
  • Cas Number : 493026-47-4
  • Molecular Weight : 244.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-hydroxymethyl-4-piperidinyl-methyl-carbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9356
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart