ADV465748429 AChemBlock

tert-Butyl 6-oxo-2-azabicyclo[2.2.1]heptane-2-carboxylate

  • MDL number MFCD13806381
  • Cas Number : 198835-04-0
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-6-oxo-2-azabicyclo-2-2-1-heptane-2-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9365
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart