ADV465748434 AChemBlock

2-Bromo-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine hydrochloride

  • MDL number MFCD08448172
  • Cas Number : 949922-52-5
  • Molecular Weight : 255.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-4-5-6-7-tetrahydrothiazolo-5-4-c-pyridine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9371
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart