ADV465748435 AChemBlock

4-(1H-Tetrazol-1-yl)phenol

  • MDL number MFCD03490124
  • Cas Number : 64001-11-2
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-1h-tetrazol-1-yl-phenol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9372
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart