ADV465748437 AChemBlock

Methyl 5H-pyrrolo[2,3-b]pyrazine-2-carboxylate

  • MDL number MFCD16658543
  • Cas Number : 1363381-89-8
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-5h-pyrrolo-2-3-b-pyrazine-2-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9374
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart