ADV465748438 AChemBlock

1-Boc-3-bromo-1H-pyrrolo[3,2-b]pyridine

  • MDL number MFCD16660027
  • Cas Number : 192189-15-4
  • Molecular Weight : 297.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-bromo-1h-pyrrolo-3-2-b-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9375
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart