ADV465748447 AChemBlock

diethyl 2-(3,5-dichlorophenyl)malonate

  • MDL number MFCD09037803
  • Cas Number : 1186194-50-2
  • Molecular Weight : 305.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/propanedioic-acid-2-3-5-dichlorophenyl-1-3-diethyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9384
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart