ADV465748448 AChemBlock

(2S,5S)-5-hydroxypiperidine-2-carboxylic acid hydrochloride

  • MDL number MFCD28166286
  • Cas Number : 154307-84-3
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2s-5s-5-hydroxypiperidine-2-carboxylic-acid-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9385
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart