ADV465748450 AChemBlock

8-Iodo-3H-pyrazolo[1,5-a][1,3,5]triazin-4-one

  • MDL number MFCD18073976
  • Cas Number : 147916-84-5
  • Molecular Weight : 262.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/8-iodo-3h-pyrazolo-1-5-a-1-3-5-triazin-4-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9387
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart