ADV465748458 AChemBlock

4-Amino-2,3-dihydro-2-methyl-1H-isoindol-1-one

  • MDL number MFCD19252207
  • Cas Number : 682757-53-5
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Physical Form Light yellow solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-amino-2-3-dihydro-2-methyl-1h-isoindol-1-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9395
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart