ADV465748461 AChemBlock

tert-butyl (4-(hydroxymethyl)-1-methylpiperidin-4-yl)carbamate

  • MDL number MFCD28166287
  • Cas Number : 1630906-32-9
  • Molecular Weight : 244.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/carbamic-acid-n-1-methyl-4-hydroxymethyl-4-piperidinyl-1-1-dimethylethyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9398
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart