ADV465748462 AChemBlock

tert-butyl 4-(3,3,3-trifluoro-2-hydroxypropyl)piperidine-1-carboxylate

  • MDL number MFCD16618527
  • Cas Number : 1228631-10-4
  • Molecular Weight : 297.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-piperidinecarboxylic-acid-4-3-3-3-trifluoro-2-hydroxypropyl-1-1-dimethylethyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9399
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart