ADV465748465 AChemBlock

(6-Methylpyridazin-3-yl)methanamine dihydrochloride

  • MDL number MFCD28155130
  • Cas Number : 1630907-25-3
  • Molecular Weight : 196.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Physical Form light yellow powder
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/6-methylpyridazin-3-yl-methanamine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9402
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart