ADV465748468 AChemBlock

Methyl (S)-4,4-difluoropyrrolidine-2-carboxylate hydrochloride

  • MDL number MFCD23106100
  • Cas Number : 156046-05-8
  • Molecular Weight : 201.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-s-4-4-difluoropyrrolidine-2-carboxylate-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9405
pictograms GHS07
hstatements H303, H313, H333
Availability details for
Quantity
  • Availability
  • Add to Cart