ADV465748469 AChemBlock

3-Bromo-5-chloro-7-azaindole

  • MDL number MFCD11100204
  • Cas Number : 866546-09-0
  • Molecular Weight : 231.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-5-chloro-7-azaindole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9406
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart