ADV465748471 AChemBlock

tert-Butyl N-(cis-3-hydroxycyclobutyl)-N-methylcarbamate

  • MDL number MFCD21362303
  • Cas Number : 1033718-10-3
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-cis-3-hydroxycyclobutyl-n-methylcarbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9408
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart