ADV465748475 AChemBlock

N-Methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine

  • MDL number MFCD01346677
  • Cas Number : 78727-16-9
  • Molecular Weight : 148.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/n-methyl-7h-pyrrolo-2-3-d-pyrimidin-4-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9412
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart