ADV465748481 AChemBlock

1-(3-Oxetanyl)-3-azetidinamine oxalate

  • MDL number MFCD28166291
  • Cas Number : 1630907-32-2
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-3-oxetanyl-3-azetidinamine-oxalate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9418
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart