ADV465748483 AChemBlock

(3S,4S)-1-Boc-pyrrolidine-3,4-diol

  • MDL number MFCD11054832
  • Cas Number : 90481-33-7
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3s-4s-1-boc-pyrrolidine-3-4-diol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9420
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart