ADV465748484 AChemBlock

Methyl 5-iodo-1H-Pyrazole-3-carboxylate

  • MDL number MFCD28166292
  • Cas Number : 1533442-31-7
  • Molecular Weight : 252.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-iodo-1h-pyrazole-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9421
pictograms GHS07
hstatements H303;H313;H333
Availability details for
Quantity
  • Availability
  • Add to Cart