ADV465748508 AChemBlock

7-Methyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine

  • MDL number MFCD28166294
  • Cas Number : 1630906-29-4
  • Molecular Weight : 137.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/7-methyl-4h-5h-6h-7h-pyrazolo-1-5-a-pyrazine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9445
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart