ADV465748510 AChemBlock

Sodium 6-chloro-[1,2,4]triazolo[4,3-b]pyridazine-3-carboxylate

  • MDL number MFCD28155131
  • Cas Number : 1630906-50-1
  • Molecular Weight : 220.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/sodium-6-chloro-1-2-4-triazolo-4-3-b-pyridazine-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9447
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart