ADV465748512 AChemBlock

7-Oxa-2-aza-spiro[4.5]decane hemioxalate

  • MDL number MFCD28166296
  • Cas Number : 1630906-43-2
  • Molecular Weight : 372.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/7-oxa-2-aza-spiro-4-5-decane-hemioxalate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9449
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart