ADV465748516 AChemBlock

trans-5-Amino-2-Boc-hexahydro-cyclopenta[c]pyrrole hydrochloride

  • MDL number MFCD28166297
  • Cas Number : 1630907-13-9
  • Molecular Weight : 262.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/trans-5-amino-2-boc-hexahydro-cyclopenta-c-pyrrole-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9453
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart