ADV465748521 AChemBlock

3-Bromo-5-fluoro-7-azaindole

  • MDL number MFCD12962622
  • Cas Number : 1190309-71-7
  • Molecular Weight : 215.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-5-fluoro-7-azaindole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9458
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart