ADV465748524 AChemBlock

3-Bromo-4,7-diaza-1H-indazole

  • MDL number MFCD09834831
  • Cas Number : 81411-68-9
  • Molecular Weight : 199.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-4-7-diaza-1h-indazole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9461
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart