ADV465748527 AChemBlock

(S)-3,3-Difluoro-cyclopentanemethanol

  • MDL number MFCD23106021
  • Cas Number : 1407991-23-4
  • Molecular Weight : 136.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/s-3-3-difluoro-cyclopentanemethanol.html

Safety Information

signalword Danger
pcodes P210;P233;P301+330+331;P303+361+353
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9464
pictograms GHS02;GHS07
hstatements H226;H302;H312;H318
Availability details for
Quantity
  • Availability
  • Add to Cart