ADV465748540 AChemBlock

5,5-Difluoropiperidin-2-one

  • MDL number MFCD18433642
  • Cas Number : 1255666-50-2
  • Molecular Weight : 135.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-5-difluoropiperidin-2-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9477
pictograms GHS07
hstatements H303, H313, H333
Availability details for
Quantity
  • Availability
  • Add to Cart