ADV465748541 AChemBlock

benzyl (S)-3-(tosyloxy)pyrrolidine-1-carboxylate

  • MDL number MFCD18252853
  • Cas Number : 158654-83-2
  • Molecular Weight : 375.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/1-pyrrolidinecarboxylic-acid-3-4-methylphenyl-sulfonyl-oxy-phenylmethyl-ester-3s.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9478
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart