ADV465748544 AChemBlock

Methyl 6-(trifluoromethyl)-4-pyrimidinecarboxylate

  • MDL number MFCD28166300
  • Cas Number : 1353101-38-8
  • Molecular Weight : 206.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-trifluoromethyl-4-pyrimidinecarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9482
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart