ADV465748545 AChemBlock

4-Boc-2-(4-(trifluoromethyl)phenyl)piperazine

  • MDL number MFCD08686043
  • Cas Number : 886767-93-7
  • Molecular Weight : 330.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/4-boc-2-4-trifluoromethyl-phenyl-piperazine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=I-9483
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart